missing translation for 'onlineSavingsMsg'
Learn More

rac N-Benzyl Nebivolol-d4, TRC

Product Code. 30328803
Change view
Click to view available options
Kvantitet:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30328803 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30328803 Supplier Toronto Research Chemicals Supplier No. TRCB28573210MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material rac N-Benzyl Nebivolol-d4
CAS 1246814-48-1
Molekylformel C29H27D4F2NO4
Kvantitet 10 mg
Synonym alpha,alpha'-[[(Phenylmethyl)imino]bis(methylene)]bis[6-fluoro-3,4-dihydro-2H-1-benzopyran-2-methanol-d4
InChI formel InChI=1S/C29H31F2NO4/c30-22-8-12-26-20(14-22)6-10-28(35-26)24(33)17-32(16-19-4-2-1-3-5-19)18-25(34)29-11-7-21-15-23(31)9-13-27(21)36-29/h1-5,8-9,12-15,24-25,28-29,33-34H,6-7,10-11,16-18H2/i17D2,18D2
LEDER [2H]C([2H])(C(O)C1CCc2cc(F)ccc2O1)N(Cc3ccccc3)C([2H])([2H])C(O)C4CCc5cc(F)ccc5O4
IUPAC-namn 2-[benzyl-[1,1-dideuterio-2-(6-fluoro-3,4-dihydro-2H-chromen-2-yl)-2-hydroxyethyl]amino]-2,2-dideuterio-1-(6-fluoro-3,4-dihydro-2H-chromen-2-yl)ethanol
Molekylvikt (g/mol) 499.58
Formel vikt 499.2472
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.