Läs mer
rac N-Bisdesmethyl Tramadol, Hydrochloride, TRC
Produktkod. 30345023
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30345023 10 mg
1 options
Produktkod. 30345023 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB42613010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material rac N-Bisdesmethyl Tramadol, Hydrochloride
CAS 541505-91-3
Molekylformel C14H22ClNO2
Kvantitet 10 mg
Synonym cis-(+/-)-2-Aminomethyl-1-(3-methoxyphenyl)cyclohexanol Hydrochloride, (1R,2R)-rel-2-(Aminomethyl)-1-(3-methoxyphenyl)-cyclohexanol Hydrochloride
InChI formel InChI=1S/C14H21NO2.ClH/c1-17-13-7-4-6-11(9-13)14(16)8-3-2-5-12(14)10-15;/h4,6-7,9,12,16H,2-3,5,8,10,15H2,1H3;1H/t12-,14+;/m1./s1
LEDER O[C@]1(C2=CC(OC)=CC=C2)CCCC[C@]1([H])CN.Cl
IUPAC-namn (1R,2R)-2-(aminomethyl)-1-(3-methoxyphenyl)cyclohexan-1-ol;hydrochloride
Molekylvikt (g/mol) 271.78