Läs mer
rac N-Desmethyl Mephenytoin, TRC
Produktkod. 30362637
Change view
Kvantitet:
5 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30362637 100 mg
30353074 5 mg
30350032 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30362637 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD292060100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 631-07-2
Molekylformel C11 H12 N2 O2
Molekylvikt (g/mol) 204.23
Synonym 2,4-Imidazolidinedione, 5-ethyl-5-phenyl-, Hydantoin, 5-ethyl-5-phenyl- (6CI,7CI,8CI), 5-Ethyl-5-phenyl-2,4-imidazolidinedione, (RS)-5-Ethyl-5-phenyl-2,4-imidazolidinedione, 5-Ethyl-5-phenylhydantoin, 5-Phenyl-5-ethylhydantoin, Desmethylmephenytoin, Ethylphenylhydantoin, NSC 150466, NSC 33388, Nirvanol, Normephenytoin
IUPAC-namn 5-ethyl-5-phenylimidazolidine-2,4-dione
LEDER CCC1(NC(=O)NC1=O)c2ccccc2
Specifikationer
Kemiskt namn eller material rac N-Desmethyl Mephenytoin
Kvantitet 100 mg
InChI formel InChI=1S/C11H12N2O2/c1-2-11(8-6-4-3-5-7-8)9(14)12-10(15)13-11/h3-7H,2H2,1H3,(H2,12,13,14,15)
Formel vikt 204.0899