Läs mer
rac-N’-Nitroso Nornicotine, TRC
Produktkod. 18051050
Change view
Kvantitet:
5 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18051050 100 mg
18070440 10 mg
18042540 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 18051050 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN535000100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 80508-23-2
Molekylformel C9 H11 N3 O
Molekylvikt (g/mol) 177.2
Synonym N-Nitrosonornicotine, rac-N'-Nitroso Nornicotine, NNN, N-Nitrosonornicotine, rac-N'-Nitroso Nornicotine
IUPAC-namn 3-(1-nitrosopyrrolidin-2-yl)pyridine
LEDER O=NN1CCCC1c2cccnc2
Specifikationer
Kemiskt namn eller material (±)-N'-Nitrosonornicotine (NNN)
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C9H11N3O/c13-11-12-6-2-4-9(12)8-3-1-5-10-7-8/h1,3,5,7,9H,2,4,6H2
Formel vikt 177.0902
Procent renhet >95%