Läs mer
rac-Naproxen 2-Propyl Ester, TRC
Produktkod. 30467607
Change view
Kvantitet:
250 mg
2.5 g
Förpackningsstorlek:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30467607 2.5 g 2.5g
30470460 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30467607 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN3775602.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 68641-85-0
Molekylformel C17H20O3
Molekylvikt (g/mol) 272.34
Synonym 6-Methoxy-alpha-methyl-2-naphthaleneacetic Acid 1-Methylethyl Ester
IUPAC-namn propan-2-yl 2-(6-methoxynaphthalen-2-yl)propanoate
LEDER COc1ccc2cc(ccc2c1)C(C)C(=O)OC(C)C
Specifikationer
Kemiskt namn eller material rac-Naproxen 2-Propyl Ester
Kvantitet 2.5 g
InChI formel InChI=1S/C17H20O3/c1-11(2)20-17(18)12(3)13-5-6-15-10-16(19-4)8-7-14(15)9-13/h5-12H,1-4H3
Formel vikt 272.1412