Läs mer
rac-Nicotine, TRC
Produktkod. 30009899
Change view
Kvantitet:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009899 100 mg
30006497 500 mg
30008204 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30009899 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN412420100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 22083-74-5
Molekylformel C10 H14 N2
Molekylvikt (g/mol) 162.23
Synonym Nicotine, racemic, (R,S)-Nicotine, 1-Methyl-2-(3-pyridyl)pyrrolidine, 3-(1-Methyl-2-pyrrolidinyl)pyridine, DL-Nicotine
IUPAC-namn 3-(1-methylpyrrolidin-2-yl)pyridine
LEDER CN1CCCC1c2cccnc2
Specifikationer
Kemiskt namn eller material rac-Nicotine
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C10H14N2/c1-12-7-3-5-10(12)9-4-2-6-11-8-9/h2,4,6,8,10H,3,5,7H2,1H3
Formel vikt 162.1157
Procent renhet >95%