Läs mer
(Rac)-NSC305787, MedChemExpress
Produktkod. 16487546
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
Förpackningsstorlek:
100mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16487546 100 mg 100mg
16480108 5 mg -
16423657 50 mg -
16407648 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16487546 Leverantör Medchem Express Leverantörsnummer HY18931
MedChemExpress NSC305787 is an inhibitor of ezrin with a Kd of 5.85 μM, inhibits the phosphorylation of ezrin caused by PKCι with an IC50 of 8.3 μM, has antitumor activity.
NSC305787 is an inhibitor of ezrin with a Kd of 5.85 μM, inhibits the phosphorylation of ezrin caused by PKCι with an IC50 of 8.3 μM, has antitumor activity.
Specifikationer
Kemiskt namn eller material (Rac)-NSC305787
Fysisk form Solid
CAS 785718-37-8
Färg Off-White
Kvantitet 100 mg
Anteckningar om renhetsgrad Research
Molekylformel C25H30Cl2N2O
För användning med (applikation) Cancer-Kinase/protease
Löslighetsinformation DMSO : 6 mg/mL (13.47 mM; Need ultrasonic and warming)
LEDER OC(C1CCCCN1)C2=CC(C3(C[C@H](C4)C5)C[C@H]5C[C@H]4C3)=NC(C(Cl)=C6)=C2C=C6Cl
Molekylvikt (g/mol) 445.42
Formel vikt 445.42
Procent renhet 98.0%