Läs mer
rac O-Methyl Tolterodine Fumarate, TRC
Produktkod. 30466027
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30466027 1 g
1 options
Produktkod. 30466027 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3308851G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material rac O-Methyl Tolterodine Fumarate
CAS 1240303-69-8
Molekylformel C27H37NO5
Kvantitet 1 g
Synonym 2-Methoxy-5-methyl-N,N-bis(1-methylethyl)-γ-phenylbenzenepropanamine (2E)-2-Butenedioate, N,N-Diisopropyl-3-(2-methoxy-5-methylphenyl)-3-phenylpropylamine Fumarate, N-[3-(2-Methoxy-5-methylphenyl)-3-phenylpropyl]diisopropylamine Fumarate, 2-[3-[Bis(1-methylethyl)amino]-1-phenyl-propyl]-4-methyl-methoxybenzene Fumarate
InChI formel InChI=1S/C23H33NO.C4H4O4/c1-17(2)24(18(3)4)15-14-21(20-10-8-7-9-11-20)22-16-19(5)12-13-23(22)25-6;5-3(6)1-2-4(7)8/h7-13,16-18,21H,14-15H2,1-6H3;1-2H,(H,5,6)(H,7,8)/b;2-1+
LEDER CC1=CC(=C(C=C1)OC)C(CCN(C(C)C)C(C)C)C2=CC=CC=C2.C(=C/C(=O)O)\C(=O)O
IUPAC-namn (E)-but-2-enedioic acid;3-(2-methoxy-5-methylphenyl)-3-phenyl-N,N-di(propan-2-yl)propan-1-amine
Molekylvikt (g/mol) 455.59
Formel vikt 455.2672