Läs mer
rac Octanoyl Carnitine Chloride, TRC
Produktkod. 30428595
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30428595 100 mg
30416552 50 mg
30426909 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30428595 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO238000100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 14919-35-8
Molekylformel C14 H28 N O4 . Cl
Molekylvikt (g/mol) 309.83
IUPAC-namn (2-carboxy-2-octanoyloxyethyl)-trimethylazanium;chloride
LEDER [Cl-].CCCCCCCC(=O)OC(C[N+](C)(C)C)C(=O)O
Specifikationer
Kemiskt namn eller material rac Octanoyl Carnitine
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C14H27NO4.ClH/c1-5-6-7-8-9-10-13(16)19-12(14(17)18)11-15(2,3)4;/h12H,5-11H2,1-4H3;1H
Formel vikt 309.1707
Procent renhet >95%