Läs mer
rac Perhexiline Maleate, TRC
Produktkod. 30428703
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30428703 25 mg
30417169 10 mg
30411982 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30428703 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP28732025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6724-53-4
Molekylformel C19 H35 N . C4 H4 O4
Molekylvikt (g/mol) 393.56
Synonym Piperidine, 2-(2,2-dicyclohexylethyl)-, (2Z)-2-butenedioate (1:1), Piperidine, 2-(2,2-dicyclohexylethyl)-, (Z)-2-butenedioate (1:1), Piperidine, 2-(2,2-dicyclohexylethyl)-, maleate (1:1) (8CI), Piperidine, 2-(2,2-dicyclohexylethyl)-, maleate (7CI), 2-(2,2-Dicyclohexylethyl)piperidine maleate, Perhexilene maleate, Perhexiline maleate, Pexid
IUPAC-namn (Z)-but-2-enedioic acid;2-(2,2-dicyclohexylethyl)piperidine
LEDER OC(=O)\C=C/C(=O)O.C(C(C1CCCCC1)C2CCCCC2)C3CCCCN3
Specifikationer
Kemiskt namn eller material rac Perhexiline maleate
Kvantitet 25 mg
InChI formel InChI=1S/C19H35N.C4H4O4/c1-3-9-16(10-4-1)19(17-11-5-2-6-12-17)15-18-13-7-8-14-20-18;5-3(6)1-2-4(7)8/h16-20H,1-15H2;1-2H,(H,5,6)(H,7,8)/b;2-1-
Formel vikt 393.2879