Läs mer
rac Ractopamine Hydrochloride (Mixture of Diastereomers), TRC
Produktkod. 30007398
Change view
Kvantitet:
50 mg
100 mg
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30007398 5 g
17974295 50 mg
17940925 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30007398 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR0714005G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 90274-24-1
Molekylformel C18 H23 N O3 . Cl H
Molekylvikt (g/mol) 337.84
Synonym 4-Hydroxy-alpha-[[[3-(4-hydroxyphenyl)-1-methylpropyl]amino]methyl]benzenemethanol, EL 737, LY 031537, LY O31537, Optaflexx, Paylean, Ractopamine hydrochloride, Topmax 9
IUPAC-namn 4-[3-[[2-hydroxy-2-(4-hydroxyphenyl)ethyl]amino]butyl]phenol;hydrochloride
LEDER Cl.CC(CCc1ccc(O)cc1)NCC(O)c2ccc(O)cc2
Specifikationer
Kemiskt namn eller material Ractopamine hydrochloride
Anteckningar om renhetsgrad HPLC
Kvantitet 5 g
InChI formel InChI=1S/C18H23NO3.ClH/c1-13(2-3-14-4-8-16(20)9-5-14)19-12-18(22)15-6-10-17(21)11-7-15;/h4-11,13,18-22H,2-3,12H2,1H3;1H
Formel vikt 337.1445
Procent renhet >95%