Läs mer
rac-Rhododendrol, TRC
Produktkod. 30420447
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30420447 10 mg
30408830 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30420447 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR31865010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material rac-Rhododendrol
CAS 69617-84-1
Molekylformel C10 H14 O2
Kvantitet 10 mg
Synonym Benzenepropanol','4-hydroxy-α-methyl-, 4-Hydroxy-α-methylbenzenepropanol, (±)-Rhododendrol, 4-(3-Hydroxybutyl)phenol, 4-(4-Hydroxyphenyl)-butan-2-ol, 4-(p-Hydroxyphenyl)-2-butanol, Betulegenol, Frambinol, NSC 40514, Rhododenol, 4-(4-Hydroxyphenyl)-2-butanol, rac-Rhododendrol, Raspberry alcohol
InChI formel InChI=1S/C10H14O2/c1-8(11)2-3-9-4-6-10(12)7-5-9/h4-8,11-12H,2-3H2,1H3
LEDER CC(O)CCc1ccc(O)cc1
IUPAC-namn 4-(3-hydroxybutyl)phenol
Molekylvikt (g/mol) 166.22
Formel vikt 166.0994