Läs mer
rac Secoisolariciresinol Diglucoside, TRC
Produktkod. 30428225
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30428225 10 mg
30413569 25 mg
30426034 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30428225 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS23951010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 158932-33-3
Molekylformel C32 H46 O16
Molekylvikt (g/mol) 686.7
Synonym Secoisolariciresinol Diglycoside, Seco-Isolariciresinol Diglycoside
IUPAC-namn (2R,3R,4S,5S,6R)-2-[(2R,3R)-2,3-bis[(4-hydroxy-3-methoxyphenyl)methyl]-4-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxybutoxy]-6-(hydroxymethyl)oxane-3,4,5-triol
LEDER COc1cc(C[C@@H](CO[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](CO[C@@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O)Cc4ccc(O)c(OC)c4)ccc1O
Specifikationer
Kemiskt namn eller material Seco-isolariciresinol diglucoside(sdg)
Kvantitet 10 mg
InChI formel InChI=1S/C32H46O16/c1-43-21-9-15(3-5-19(21)35)7-17(13-45-31-29(41)27(39)25(37)23(11-33)47-31)18(8-16-4-6-20(36)22(10-16)44-2)14-46-32-30(42)28(40)26(38)24(12-34)48-32/h3-6,9-10,17-18,23-42H,7-8,11-14H2,1-2H3/t17-,18-,23+,24+,25+,26+,27-,28-,29+,30+,31+,32+/m0/s1
Formel vikt 686.2786