Läs mer
rac Secoisolariciresinol, TRC
Produktkod. 30427686
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30427686 1 mg
1 options
Produktkod. 30427686 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS2395001MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material rac Secoisolariciresinol
CAS 75365-01-4
Molekylformel C20H26O6
Kvantitet 1 mg
Synonym (2R,3R)-rel-2,3-Bis[(4-hydroxy-3-methoxyphenyl)methyl]-1,4-butanediol, (R*,R*)-(±)-2,3-Bis[(4-hydroxy-3-methoxyphenyl)methyl]-1,4-butanediol, (R*,R*)-2,3-Bis[(4-hydroxy-3-methoxyphenyl)methyl]-1,4-butanediol
InChI formel InChI=1S/C20H26O6/c1-25-19-9-13(3-5-17(19)23)7-15(11-21)16(12-22)8-14-4-6-18(24)20(10-14)26-2/h3-6,9-10,15-16,21-24H,7-8,11-12H2,1-2H3/t15-,16-/m1/s1
LEDER COC1=C(C=CC(=C1)C[C@H](CO)[C@H](CC2=CC(=C(C=C2)O)OC)CO)O
Molekylvikt (g/mol) 362.42