Läs mer
rac-Synephrine, TRC
Produktkod. 30011109
Change view
Kvantitet:
50 mg
100 mg
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30011109 100 mg
30006443 5 g
30009614 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30011109 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS920000100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 94-07-5
Molekylformel C9 H13 N O2
Molekylvikt (g/mol) 167.2
Synonym Benzenemethanol, 4-hydroxy-α-[(methylamino)methyl]-, Benzyl alcohol, p-hydroxy-α-[(methylamino)methyl]- (8CI), (±)-N-Methyloctopamine, (±)-S38537-9, (±)-Synephrine, 1-(4-Hydroxyphenyl)-2-(methylamino)ethanol, 1-(4-Hydroxyphenyl)-N-methylethanolamine, 1-Hydroxy-4-[(1-hydroxy-2-methylamino)ethyl]benzene, Advantra Z, Analeptin, DL-Synephrine, Ethaphene, N-[2-Hydroxy-2-(4-hydroxyphenyl)ethyl]-N-methylamine, NSC 166285, NSC 170956, Oxedrine, Parasympatol, S 38537-9, Simpalon, Simpatol, Sympaethamin, Sympaethamine, Sympathol, Synephrin, Synephrine, Synthenate, dl-1-(4-Hydroxyphenyl)-2-(methylamino)ethanol, p-Hydroxy-α-[(methylamino)methyl]benzyl alcohol, β-Methylamino-α-(4-hydroxyphenyl)ethyl alcohol
IUPAC-namn 4-[1-hydroxy-2-(methylamino)ethyl]phenol
LEDER CNCC(O)c1ccc(O)cc1
Specifikationer
Kemiskt namn eller material Synephrine
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C9H13NO2/c1-10-6-9(12)7-2-4-8(11)5-3-7/h2-5,9-12H,6H2,1H3
Formel vikt 167.0946
Procent renhet >95%