Läs mer
rac Tamsulosin Hydrochloride, TRC
Produktkod. 30136003
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136003 25 mg
30135147 100 mg
30008015 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136003 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT00635125MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 80223-99-0
Molekylformel C20 H28 N2 O5 S . Cl H
Molekylvikt (g/mol) 444.973
Synonym 5-[(2RS)-2-[[2-(2-Ethoxyphenoxy)ethyl]amino]propyl]-2-methoxybenzenesulfonamide Hydrochloride, Tamsulosin Hydrochloride Imp. G (EP) as Racemate Hydrochloride, Tamsulosin Racemate Hydrochloride, Benzenesulfonamide, 5-[2-[[2-(2-ethoxyphenoxy)ethyl]amino]propyl]-2-methoxy-, hydrochloride (1:1), Benzenesulfonamide, 5-[2-[[2-(2-ethoxyphenoxy)ethyl]amino]propyl]-2-methoxy-, monohydrochloride (9CI), LY 253352, YM 12617, dl-Tamsulosin hydrochloride
IUPAC-namn 5-[2-[2-(2-ethoxyphenoxy)ethylamino]propyl]-2-methoxybenzenesulfonamide;hydrochloride
LEDER Cl.CCOc1ccccc1OCCNC(C)Cc2ccc(OC)c(c2)S(=O)(=O)N
Specifikationer
Kemiskt namn eller material 5-[(2RS)-2-[(2-(2-Ethoxyphenoxy)ethyl)amino]propyl]-2-methoxybenzenesulfonamide Hydrochloride (Tamsulosin Racemate Hydrochloride)
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C20H28N2O5S.ClH/c1-4-26-17-7-5-6-8-18(17)27-12-11-22-15(2)13-16-9-10-19(25-3)20(14-16)28(21,23)24;/h5-10,14-15,22H,4,11-13H2,1-3H3,(H2,21,23,24);1H
Formel vikt 444.149
Procent renhet >95%