Läs mer
rac threo-9,10-Dihydroxystearic Acid, TRC
Produktkod. 30373176
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30373176 100 mg
1 options
Produktkod. 30373176 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD454750100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material rac threo-9,10-Dihydroxystearic Acid
CAS 2391-05-1
Molekylformel C18 H36 O4
Kvantitet 100 mg
Synonym Octadecanoic acid, 9,10-dihydroxy-, (9R,10R)-rel-, Octadecanoic acid, 9,10-dihydroxy-, (R*,R*)-, Octadecanoic acid, 9,10-dihydroxy-, threo- (8CI), rel-(9R,10R)-9,10-Dihydroxyoctadecanoic acid, threo-9,10-Dihydroxyoctadecanoic acid, threo-9,10-Dihydroxystearic acid, trans-9,10-Dihydroxystearic acid
InChI formel InChI=1S/C18H36O4/c1-2-3-4-5-7-10-13-16(19)17(20)14-11-8-6-9-12-15-18(21)22/h16-17,19-20H,2-15H2,1H3,(H,21,22)/t16-,17-/m0/s1
LEDER CCCCCCCC[C@H](O)[C@@H](O)CCCCCCCC(=O)O
IUPAC-namn (9S,10S)-9,10-dihydroxyoctadecanoic acid
Molekylvikt (g/mol) 316.48
Formel vikt 316.2614