Läs mer
rac-trans 3’-Hydroxymethylnicotine Hemisuccinate N-Hydroxysuccinimide Ester, TRC
Produktkod. 30404053
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30404053 5 mg
1 options
Produktkod. 30404053 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH9481955MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material rac-trans 3'-Hydroxymethylnicotine Hemisuccinate N-Hydroxysuccinimide Ester
CAS 1207282-60-7
Molekylformel C19H23N3O6
Kvantitet 5 mg
Synonym Rac-Trans 3'-Hydroxymethylnicotine Hemisuccinate N-Hydroxysuccinimide Ester
InChI formel InChI=1S/C19H23N3O6/c1-21-10-8-14(19(21)13-3-2-9-20-11-13)12-27-17(25)6-7-18(26)28-22-15(23)4-5-16(22)24/h2-3,9,11,14,19H,4-8,10,12H2,1H3/t14-,19-/m1/s1
LEDER CN1CC[C@@H]([C@H]1C2=CN=CC=C2)COC(=O)CCC(=O)ON3C(=O)CCC3=O
Molekylvikt (g/mol) 389.4
Formel vikt 389.1587