Läs mer
rac-trans 3’-Thiomethyl Nicotine Dihydrochloride, TRC
Produktkod. 30449382
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30449382 5 mg
30449209 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30449382 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT3525005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1246820-13-2
Molekylformel C11H16N2S • 2(HCl)
Molekylvikt (g/mol) 280.06
IUPAC-namn ((2S,3S)-1-methyl-2-(pyridin-3-yl)pyrrolidin-3-yl)methanethiol dihydrochloride
LEDER CN1[C@H](C2=CC=CN=C2)[C@@H](CS)CC1.Cl.Cl
Specifikationer
Kemiskt namn eller material rac-trans 3'-Thiomethyl Nicotine Dihydrochloride
Kvantitet 5 mg
InChI formel InChI=1S/C11H16N2S.2ClH/c1-13-6-4-10(8-14)11(13)9-3-2-5-12-7-9;;/h2-3,5,7,10-11,14H,4,6,8H2,1H3;2*1H/t10-,11-;;/m1../s1
Formel vikt 280.0568