Läs mer
rac trans-4-Cotinine Carboxylic Acid Methyl Ester, TRC
Produktkod. 30357831
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30357831 500 mg
30356332 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30357831 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC725155500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material rac trans-4-Cotinine Carboxylic Acid Methyl Ester
CAS 135028-97-6
Molekylformel C12H14N2O3
Kvantitet 500 mg
Synonym (2R,3R)-rel-1-Methyl-5-oxo-2-(3-pyridinyl)-3-pyrrolidinecarboxylic Acid Methyl Ester, trans-1-Methyl-5-oxo-2-(3-pyridinyl)-3-pyrrolidinecarboxylic Acid Methyl Ester
InChI formel InChI=1S/C12H14N2O3/c1-14-10(15)6-9(12(16)17-2)11(14)8-4-3-5-13-7-8/h3-5,7,9,11H,6H2,1-2H3/t9-,11-/m0/s1
LEDER COC(=O)[C@H]1CC(=O)N(C)[C@H]1c2cccnc2
IUPAC-namn methyl (2R,3S)-1-methyl-5-oxo-2-pyridin-3-ylpyrrolidine-3-carboxylate
Molekylvikt (g/mol) 234.25
Formel vikt 234.1004