Läs mer
rac trans-9,10-Epoxystearic Acid, TRC
Produktkod. 30138100
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138100 25 mg
30011930 5 mg
30007548 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30138100 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE58990025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 13980-07-9
Molekylformel C18 H34 O3
Molekylvikt (g/mol) 298.46
IUPAC-namn 8-[(2R,3R)-3-octyloxiran-2-yl]octanoic acid
LEDER CCCCCCCC[C@H]1O[C@@H]1CCCCCCCC(=O)O
Specifikationer
Kemiskt namn eller material trans-9,10-Epoxystearic Acid
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C18H34O3/c1-2-3-4-5-7-10-13-16-17(21-16)14-11-8-6-9-12-15-18(19)20/h16-17H,2-15H2,1H3,(H,19,20)/t16-,17-/m1/s1
Formel vikt 298.2508
Procent renhet >95%