Läs mer
rac-trans-Ketoconazole, TRC
Produktkod. 30134156
Change view
Kvantitet:
2.5 mg
10 mg
25 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30134156 10 mg -
30006164 25 mg -
18052040 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134156 Leverantör Toronto Research Chemicals Leverantörsnummer TRCK18601010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 83374-59-8
Molekylformel C26 H28 Cl2 N4 O4
Molekylvikt (g/mol) 531.43
Synonym Piperazine, 1-acetyl-4-[4-[[(2R,4R)-2-(2,4-dichlorophenyl)-2-(1H-imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]-, rel-, Piperazine, 1-acetyl-4-[4-[[2-(2,4-dichlorophenyl)-2-(1H-imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]-, trans-, rel-1-Acetyl-4-[4-[[(2R,4R)-2-(2,4-dichlorophenyl)-2-(1H-imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazine, R 44319, RS 36745, trans-Ketoconazole, 1-Acetyl-4-[4-[[(2RS,4RS)-2-(2,4-dichlorophenyl)-2-(1H-imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazine, Ketoconazole EP impurity C
IUPAC-namn 1-[4-[4-[[(2R,4R)-2-(2,4-dichlorophenyl)-2-(imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazin-1-yl]ethanone
LEDER CC(=O)N1CCN(CC1)c2ccc(OC[C@@H]3CO[C@@](Cn4ccnc4)(O3)c5ccc(Cl)cc5Cl)cc2
Specifikationer
Kemiskt namn eller material trans-Ketoconazole
Kvantitet 10 mg
InChI formel InChI=1S/C26H28Cl2N4O4/c1-19(33)31-10-12-32(13-11-31)21-3-5-22(6-4-21)34-15-23-16-35-26(36-23,17-30-9-8-29-18-30)24-7-2-20(27)14-25(24)28/h2-9,14,18,23H,10-13,15-17H2,1H3/t23-,26+/m1/s1
Formel vikt 530.1488