Läs mer
rac-trans-N-Methyl Sertraline Hydrochloride, TRC
Produktkod. 30475621
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30475621 10 mg
1 options
Produktkod. 30475621 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM32647010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material rac-trans-N-Methyl Sertraline Hydrochloride
CAS 79836-76-3
Molekylformel C18H20Cl3N
Kvantitet 10 mg
Synonym (1R,4S)-rel-4-(3,4-Dichlorophenyl)-1,2,3,4-tetrahydro-N,N-dimethyl-1-naphthalenamine Hydrochloride, trans-4-(3,4-Dichlorophenyl)-1,2,3,4-tetrahydro-N,N-dimethyl-1-naphthalenamine Hydrochloride, trans-(±)-4-(3,4-Dichlorophenyl)-1,2,3,4-tetrahydro-N,N-dimethyl-1-naphthalenamine Hydrochloride
InChI formel InChI=1S/C18H19Cl2N.ClH/c1-21(2)18-10-8-13(14-5-3-4-6-15(14)18)12-7-9-16(19)17(20)11-12;/h3-7,9,11,13,18H,8,10H2,1-2H3;1H/t13-,18+;/m0./s1
LEDER Cl.CN(C)[C@@H]1CC[C@@H](c2ccc(Cl)c(Cl)c2)c3ccccc13
IUPAC-namn (1R,4S)-4-(3,4-dichlorophenyl)-N,N-dimethyl-1,2,3,4-tetrahydronaphthalen-1-amine;hydrochloride
Molekylvikt (g/mol) 356.72
Formel vikt 355.0661