Läs mer
rac-Vestitone, TRC
Produktkod. 30456338
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30456338 100 mg
30448962 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30456338 Leverantör Toronto Research Chemicals Leverantörsnummer TRCV128900100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 57462-46-1
Molekylformel C16H14O5
Molekylvikt (g/mol) 286.28
Synonym 2,3-Dihydro-7-hydroxy-3-(2-hydroxy-4-methoxyphenyl)-4H-1-benzopyran-4-one, (±)-2',7-Dihydroxy-4'-methoxyisoflavanone, (±)-Vestitone
IUPAC-namn 7-hydroxy-3-(2-hydroxy-4-methoxyphenyl)-2,3-dihydrochromen-4-one
LEDER OC1=CC(OC)=CC=C1C(CO2)C(C3=C2C=C(O)C=C3)=O
Specifikationer
Kemiskt namn eller material rac-Vestitone
Kvantitet 100 mg
InChI formel InChI=1S/C16H14O5/c1-20-10-3-5-11(14(18)7-10)13-8-21-15-6-9(17)2-4-12(15)16(13)19/h2-7,13,17-18H,8H2,1H3
Formel vikt 286.0841