Läs mer
rac Viloxazine Hydrochloride, TRC
Produktkod. 30455538
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30455538 10 mg
1 options
Produktkod. 30455538 Leverantör Toronto Research Chemicals Leverantörsnummer TRCV31244010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Viloxazine Hydrochloride
CAS 35604-67-2
Molekylformel C13 H19 N O3 . Cl H
Kvantitet 10 mg
Synonym Morpholine','2-[(2-ethoxyphenoxy)methyl]-','hydrochloride (1:1), Morpholine','2-[(2-ethoxyphenoxy)methyl]-','hydrochloride (9CI), 2-[(2-Ethoxyphenoxy)methyl]morpholinium chloride, Catatrol, ICI 58834, Vicilan, Viloxazin hydrochloride, Viloxazine hydrochloride, Vivalan, Vivarint, Vivilan
InChI formel InChI=1S/C13H19NO3.ClH/c1-2-15-12-5-3-4-6-13(12)17-10-11-9-14-7-8-16-11;/h3-6,11,14H,2,7-10H2,1H3;1H
LEDER Cl.CCOc1ccccc1OCC2CNCCO2
IUPAC-namn 2-[(2-ethoxyphenoxy)methyl]morpholine;hydrochloride
Molekylvikt (g/mol) 273.76
Formel vikt 273.1132