Läs mer
rac Viloxazine Hydrochloride, TRC
Produktkod. 30132752
Change view
Kvantitet:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30132752 500 mg
30009593 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30132752 Leverantör Toronto Research Chemicals Leverantörsnummer TRCV312440500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 35604-67-2
Molekylformel C13 H19 N O3 . Cl H
Molekylvikt (g/mol) 273.76
Synonym Morpholine, 2-[(2-ethoxyphenoxy)methyl]-, hydrochloride (1:1), Morpholine, 2-[(2-ethoxyphenoxy)methyl]-, hydrochloride (9CI), 2-[(2-Ethoxyphenoxy)methyl]morpholinium chloride, Catatrol, ICI 58834, Vicilan, Viloxazin hydrochloride, Viloxazine hydrochloride, Vivalan, Vivarint, Vivilan
IUPAC-namn 2-[(2-ethoxyphenoxy)methyl]morpholine;hydrochloride
LEDER Cl.CCOc1ccccc1OCC2CNCCO2
Specifikationer
Kemiskt namn eller material Viloxazine Hydrochloride
Kvantitet 500 mg
InChI formel InChI=1S/C13H19NO3.ClH/c1-2-15-12-5-3-4-6-13(12)17-10-11-9-14-7-8-16-11;/h3-6,11,14H,2,7-10H2,1H3;1H
Formel vikt 273.1132