missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rac1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24072-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a GTPase which belongs to the RAS superfamily of small GTP-binding proteins. Members of this superfamily appear to regulate a diverse array of cellular events, including the control of cell growth, cytoskeletal reorganization, and the activation of protein kinases. Two transcript variants encoding different isoforms have been found for this gene.Specifications
Rac1 | |
Polyclonal | |
Unconjugated | |
RAC1 | |
MIG5, p21 Rac1, RAC1, RAC1b, Ras like protein TC25, TC 25, TC25 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
19353, 363875, 5879 | |
-20°C | |
Liquid |
Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P63000, P63001, Q6RUV5 | |
Rac1 | |
Rac1 Fusion Protein Ag17779 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |