missing translation for 'onlineSavingsMsg'
Läs mer

Racanisodamine, Dr. Ehrenstorfer

Product Code. 30312990
Change view
Click to view available options
Kvantitet:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30312990 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30312990 Supplier Dr. Ehrenstorfer Supplier No. DREC16804300

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Racanisodamine
CAS 17659-49-3
Analyt- eller komponentnamn Racanisodamine
Frakt skick +20°C
Molekylformel C17 H23 N O4
Certifikat och efterlevnad ISO/IEC 17025
Synonym α-(Hydroxymethyl)-benzeneacetic acid 6-hydroxy-8-methyl-8-azabicyclo[3.2.1]oct-3-yl ester, Tropic acid, 6-hydroxy-3-tropanyl ester (6CI), 6-Hydroxy-8-methyl-8-azabicyclo[3.2.1]oct-3-yl 3-hydroxy-2-phenylpropanoate, Racanisodamine
InChI formel InChI=1S/C17H23NO4/c1-18-12-7-13(9-15(18)16(20)8-12)22-17(21)14(10-19)11-5-3-2-4-6-11/h2-6,12-16,19-20H,7-10H2,1H3
LEDER CN1C2CC(CC1C(O)C2)OC(=O)C(CO)c3ccccc3
IUPAC-namn (6-hydroxy-8-methyl-8-azabicyclo[3.2.1]octan-3-yl) 3-hydroxy-2-phenylpropanoate
Molekylvikt (g/mol) 305.3688
Kvantitet 50 mg
Formel vikt 305.1627 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.