missing translation for 'onlineSavingsMsg'
Learn More

Tenak™ Standard Rack for Microtiterplates, for Chest Freezers, 45 mm Shelf Height

Stainlesss Steel Standard Rack for chest freezers is designed to hold microtiterplates.

Brand:  Tenak™ TE27170

Additional Details : Weight : 1.15000kg

 View more versions of this product

Product Code. 15220894

Quantity Price
1 964.00 SEK / Each
10 874.54 SEK / Each
Estimated Shipment: 16-09-2021

Add to basket


Description

Description

  • For use in chest freezers
  • Racks come with a locking rod to secure the cryoboxes
  • Constructed of stainless steel
Specifications

Specifications

135 x 92 x 516 mm
45 mm
Locking rod
Chest Frseezers
Microtiterplates (11)
Stainless Steel
Videos
SDS
Documents
Certificates