missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tenak™ Freezer Rack for 144 Matrix Plates
Freezer rack designed to hold matrix plates.
Brand: Tenak™ TE27705
Additional Details : Weight : 4.40000kg
Description
- Rack
- For use with freezers
Specifications
Matrix Plates (144) | |
Freezers | |
60 x 20 x 120 mm | |
54 mm |