Learn More
Tenak™ Classic Racks for Chest Freezers
Providing safe and secure cold storage. Tenak™ Classic Racks for Chest Freezers can be used with all freezer models.
Brand: Tenak™ TE21357
Additional Details : Weight : 1.90000kg
Description
- Racks come with a locking rod to secure cryoboxes
- Manufactured from stainless steel
- Sold without cryoboxes
Specifications
Freezer Rack | |
Stainless Steel | |
1 Pack | |
140 mm | |
32 mm |
Boxes (15) | |
Chest freezers | |
140 x 140 x 540mm | |
540 mm | |
140 mm |