missing translation for 'onlineSavingsMsg'
Learn More

Tenak™ Standard Rack for Microtiterplates, for Upright Freezers, 85 mm Shelf Height

Stainlesss Steel Standard Rack for use in upright freezers is designed to hold microtiterplates.

Brand:  Tenak™ TE27610

Additional Details : Weight : 2.40000kg

 View more versions of this product

Product Code. 15210994

Quantity Price
1 1130.00 SEK / Each
10 1024.23 SEK / Each
Estimated Shipment: 07-09-2021

Add to basket


Description

Description

  • For use in upright freezers
  • Racks can either be used as one plate per shelf or the plates can be stacked
  • Constructed of stainless steel
Specifications

Specifications

2 x 7
Upright Freezers
Microtiterplates (14)
630 x 135 x 173 mm
85 mm
Stainless Steel
Videos
SDS
Documents
Certificates