missing translation for 'onlineSavingsMsg'
Learn More

Tenak™ Standard Rack for Upright Freezer, 75 mm Box Height, with Boxes

Standard Rack fits into any upright freezer and can be used with all standard cryoboxes.

Brand:  Tenak™ TE23174B

Additional Details : Weight : 3.62000kg

 View more versions of this product

Product Code. 15220684

Quantity Price
1 1507.00 SEK / Each
10 1368.06 SEK / Each
Estimated Shipment: 21-07-2021

Add to basket


Description

Description

  • Measure the inside height and depth of your freezer when choosing the right rack
  • Racks can be used with all freezer brands
  • Constructed of stainless steel
  • Includes standard white cardboard boxes with 9 x 9 cell dividers
Specifications

Specifications

4 x 3
Upright Freezers
Boxes (12)
Stainless Steel
422 x 139 x 320 mm
75 mm (Box)
White cardboard boxes, 9 x 9 cell dividers
Videos
SDS
Documents
Certificates