Learn More
Tenak™ Standard Rack for Upright Freezer, 50 mm Box Height, with Boxes
Standard Rack fits into any upright freezer and can be used with all standard cryoboxes.
Brand:  Tenak™ TE23154B
Additional Details : Weight : 5.96000kg
 View more versions of this product
Product Code. 15220674
  • 2400.00 SEK / Each
Estimated Shipment: 04-07-2024
to see stock.
Add to basket
Description
Description
  • Measure the inside height and depth of your freezer when choosing the right rack
  • Racks can be used with all freezer brands
  • Constructed of stainless steel
  • Includes standard white cardboard boxes with 9 x 9 cell dividers
Specifications
Specifications
White cardboard boxes, 9 x 9 cell dividers
Boxes (25)
Upright Freezers
50 mm (Box)
5 x 5
Stainless Steel
683 x 139 x 276 mm