missing translation for 'onlineSavingsMsg'
Learn More

Tenak™ Comfort Rack for Upright Freezers, 100 mm Box Height, with Boxes

Comfort Rack designed to ensure a more ergonomical operating process when working with samples.

Brand:  Tenak™ TE24304B

1378.43 SEK valid until 2021-07-31
Use promo code "16115" to get your promotional price.



Additional Details : Weight : 3.77100kg

 View more versions of this product

Product Code. 15230784

Quantity Price
1 2411.00 SEK / Each
6 2190.64 SEK / Each
In stock, ready to ship: 30

Add to basket


Description

Description

  • In the comfort rack, the cryoboxes are placed in drawers
  • Increases the flexibility and reduces the operating weight compared to the rack types
  • Measure the inside height and depth of you freezer when choosing the right rack
  • Racks can be used with all freezer models
  • Constructed of stainless steel
  • Include standard white cardboard boxes with 9 x 9 cell dividers
Specifications

Specifications

3 x 4
Upright Freezers
Stainless Steel
562 x 140 x 315 mm
Boxes (12)
100 mm (Box)
Videos
SDS
Documents
Certificates