Learn More
Tenak™ Microtiterplates Comfort Rack For Upright Freezers
Rack designed to hold microtiterplates in upright freezers.
Brand: Tenak™ TE35539
Additional Details : Weight : 0.55000kg
Description
- Variety of shelf heights available
- Racks can either be used as one plate per shelf or the plates can be stacked
- Includes an alert preventing the shelves from sliding out
Specifications
Microtiterplate Rack | |
Plates (90) | |
25 mm |
15 x 6 | |
Upright Freezers |