Läs mer
Ractopamine-10-O-beta-glucuronide (Mixture of Diastereomers), TRC
Produktkod. 30425204
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30425204 5 mg
1 options
Produktkod. 30425204 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR0713955MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ractopamine-10-O-beta-glucuronide (Mixture of Diastereomers)
CAS 166022-10-2
Molekylformel C24 H31 N O9
Kvantitet 5 mg
Synonym 4-[3-[[2-Hydroxy-2-(4-hydroxyphenyl)ethyl]amino]butyl]phenyl beta-D-Glucopyranosiduronic Acid, (2S,3S,4S,5R,6S)-3,4,5-Trihydroxy-6-(4-(3-((2-hydroxy-2-(4-hydroxyphenyl)ethyl)amino)butyl)phenoxy)tetrahydro-2H-pyran-2-carboxylic Acid
InChI formel InChI=1S/C24H31NO9/c1-13(25-12-18(27)15-6-8-16(26)9-7-15)2-3-14-4-10-17(11-5-14)33-24-21(30)19(28)20(29)22(34-24)23(31)32/h4-11,13,18-22,24-30H,2-3,12H2,1H3,(H,31,32)/t13?,18?,19-,20-,21+,22-,24+/m0/s1
LEDER CC(CCc1ccc(O[C@@H]2O[C@@H]([C@@H](O)[C@H](O)[C@H]2O)C(=O)O)cc1)NCC(O)c3ccc(O)cc3
IUPAC-namn (2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-[4-[3-[[2-hydroxy-2-(4-hydroxyphenyl)ethyl]amino]butyl]phenoxy]oxane-2-carboxylic acid
Molekylvikt (g/mol) 477.504
Formel vikt 477.2