Läs mer
Ractopamine Methyl Ether, TRC
Produktkod. 30415061
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30415061 10 mg
30411354 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30415061 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR07143010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1329610-95-8
Molekylformel C19H25NO3
Molekylvikt (g/mol) 315.41
Synonym 4-Hydroxy-alpha-[[[3-(4-hydroxyphenyl)-1-methylpropyl]amino]methyl]benzenemethoxymethanol, O-Methyl-optaflexx, O-Methyl-paylean, O-Methyl-topmax 9, O-Methyl Ractopamine
IUPAC-namn 4-[3-[[2-(4-hydroxyphenyl)-2-methoxyethyl]amino]butyl]phenol
LEDER COC(CNC(C)CCc1ccc(O)cc1)c2ccc(O)cc2
Specifikationer
Kemiskt namn eller material Ractopamine Methyl Ether
Kvantitet 10 mg
InChI formel InChI=1S/C19H25NO3/c1-14(3-4-15-5-9-17(21)10-6-15)20-13-19(23-2)16-7-11-18(22)12-8-16/h5-12,14,19-22H,3-4,13H2,1-2H3
Formel vikt 315.1834