missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAE1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20491-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Mutations in the Schizosaccharomyces pombe Rae1 and Saccharomyces cerevisiae Gle2 genes have been shown to result in accumulation of poly-containing mRNA in the nucleus, suggesting that the encoded proteins are involved in RNA export. The protein encoded by this gene is a homolog of yeast Rae1. It contains four WD40 motifs, and has been shown to localize to distinct foci in the nucleoplasm, to the nuclear rim, and to meshwork-like structures throughout the cytoplasm. This gene is thought to be involved in nucleocytoplasmic transport, and in directly or indirectly attaching cytoplasmic mRNPs to the cytoskeleton. Alternatively spliced transcript variants encoding the same protein have been found for this gene.Specifications
RAE1 | |
Polyclonal | |
Unconjugated | |
RAE1 | |
MIG14, Mnrp41, mRNA export factor, MRNP41, RAE1, Rae1 protein homolog | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
8480 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P78406 | |
RAE1 | |
RAE1 Fusion Protein Ag12102 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |