Läs mer
Rafoxanide, TRC
Produktkod. 17911615
Change view
Kvantitet:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17911615 500 mg
17932265 1 g
17963305 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17911615 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR073500500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 22662-39-1
Molekylformel C19 H11 Cl2 I2 N O3
Molekylvikt (g/mol) 626.01
Synonym Benzamide, N-[3-chloro-4-(4-chlorophenoxy)phenyl]-2-hydroxy-3,5-diiodo-, Salicylanilide, 3'-chloro-4'-(p-chlorophenoxy)-3,5-diiodo- (8CI), 3,5-Diiodo-3'-chloro-4'-(p-chlorophenoxy)salicylanilide, 3,5-Diodo-3'-chloro-4'-(p-chlorophenoxy)salicylanilide, 3'-Chloro-4'-(4-chlorophenoxy)-3,5-diiodosalicylanilide, 3'-Chloro-4'-(p-chlorophenoxy)-3,5-diiodosalicylanilide, Disalan, NSC 355278, Rafoxanide, Ranide
IUPAC-namn N-[3-chloro-4-(4-chlorophenoxy)phenyl]-2-hydroxy-3,5-diiodobenzamide
LEDER Oc1c(I)cc(I)cc1C(=O)Nc2ccc(Oc3ccc(Cl)cc3)c(Cl)c2
Specifikationer
Kemiskt namn eller material Rafoxanide
Anteckningar om renhetsgrad HPLC
Kvantitet 500 mg
InChI formel InChI=1S/C19H11Cl2I2NO3/c20-10-1-4-13(5-2-10)27-17-6-3-12(9-15(17)21)24-19(26)14-7-11(22)8-16(23)18(14)25/h1-9,25H,(H,24,26)
Formel vikt 624.8205
Procent renhet >95%