Läs mer
Tocris Bioscience™ RAGE 229
Beskrivning
RAGE 229 is an antagonist of the interaction between the cytoplasmic tail of the receptor for advanced glycation end products (ctRAGE) and the formin, Diaphanous-1 (DIAPH1) (KD for binding to ctRAGE = 2 nM). RAGE 229 inhibits the migration of human aortic smooth muscle cells in an in vitro wound healing assay (IC50 = 120 nM). RAGE 229 reduces short- and long-term complications of diabetes in mouse models, without lowering blood glucose concentrations. RAGE 229 also reduces plasma concentrations of TNF-α, IL-6, and CCL2/JE-MCP1 and attenuates inflammatory signaling in diabetic mice.
Specifikationer
Specifikationer
| Kemiskt namn eller material | N-[4-[7-Cyano-4-(4-morpholinylmethyl)-2-quinolinyl]phenyl]acetamide |
| CAS | 2143072-85-7 |
| Kvantitet | 5 mg |
| Mål | AGER |
| Molekylformel | C23H22N4O2 |
| Renhet | 0.98 |