missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAI14 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17507-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
RAI14 plays a role in actin regulation at the ectoplasmic specialization, a type of cell junction specific to testis. Important for establishment of sperm polarity and normal spermatid adhesion. May also promote integrity of Sertoli cell tight junctions at the blood-testis barrier.Specifications
RAI14 | |
Polyclonal | |
Unconjugated | |
RAI14 | |
Ankycorbin, KIAA1334, NORPEG, RAI13, RAI14, retinoic acid induced 14 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26064, 294804, 75646 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5U312, Q9EP71, Q9P0K7 | |
Rai14 | |
RAI14 Fusion Protein Ag11708 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |