Learn More
Mettler Toledo™ Rainbow Bottle Set
Product Code. 15361034
Change view
Click to view available options
Packaging:
6 x 250 mL
Unit Size:
1 set
Product Code. Packaging Unit Size
15361034 6 x 250 mL 1 set
1 options
Product Code. 15361034 Supplier Mettler Toledo™ Supplier No. 30095312
  • ONLINE EXCLUSIVE PRICE
    1335.00 SEK
    1179.52 SEK / 1 uppsättningar
    Save 155.48 SEK
    11% Av
View all promotions
I lager
The pH Value at 25 °C -> 4.0, 1.7, 7.0, 9.2 | Standard measurement uncertainty -> ±0.02 pH
  • Laboratory pH Buffer
  • pH 4.01/7.00/9.21
  • Uncertainty ±0.02 pH
TRUSTED_SUSTAINABILITY
Specifications
Description Bottle Rainbow Kit 1, 6x250mL
Product Line Rainbow Bottle Set
Packaging 6 x 250 mL