Läs mer
Ralfinamide Mesylate, TRC
Produktkod. 30416180
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30416180 25 mg
1 options
Produktkod. 30416180 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR07500025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ralfinamide Mesylate
CAS 202825-45-4
Molekylformel C17 H19 F N2 O2 . C H4 O3 S
Kvantitet 25 mg
Synonym (2S)-2-[[[4-[(2-Fluorophenyl)methoxy]phenyl]methyl]amino]propanamide Methanesulfonate, (S)-Monomethanesulfonate, (S)-2-[[4-[(2-Fluorobenzyl)oxy]benzyl]amino]propanamide Methanesulfonate
InChI formel InChI=1S/C17H19FN2O2.CH4O3S/c1-12(17(19)21)20-10-13-6-8-15(9-7-13)22-11-14-4-2-3-5-16(14)18;1-5(2,3)4/h2-9,12,20H,10-11H2,1H3,(H2,19,21);1H3,(H,2,3,4)/t12-;/m0./s1
LEDER C[C@H](NCc1ccc(OCc2ccccc2F)cc1)C(=O)N.CS(=O)(=O)O
IUPAC-namn (2S)-2-[[4-[(2-fluorophenyl)methoxy]phenyl]methylamino]propanamide;methanesulfonic acid
Molekylvikt (g/mol) 398.449
Formel vikt 398.1312