Läs mer
Raloxifene 6-Glucuronide, TRC
Produktkod. 30425626
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30425626 1 mg
30420834 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30425626 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR1000251MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 174264-50-7
Molekylformel C34H35NO10S
Molekylvikt (g/mol) 649.71
Synonym Raloxifene-6-Glucuronide
LEDER O[C@@H]1[C@@H](O)[C@H](Oc2ccc3c(C(=O)c4ccc(OCCN5CCCCC5)cc4)c(sc3c2)c6ccc(O)cc6)O[C@@H]([C@H]1O)C(=O)O
Specifikationer
Kemiskt namn eller material Raloxifene 6-Glucuronide
Anteckningar om renhetsgrad HPLC
Kvantitet 1 mg
InChI formel InChI=1S/C34H35NO10S/c36-21-8-4-20(5-9-21)32-26(27(37)19-6-10-22(11-7-19)43-17-16-35-14-2-1-3-15-35)24-13-12-23(18-25(24)46-32)44-34-30(40)28(38)29(39)31(45-34)33(41)42/h4-13,18,28-31,34,36,38-40H,1-3,14-17H2,(H,41,42)/t28-,29-,30+,31-,34+/m0/s1
Formel vikt 649.1982
Procent renhet >95%