Läs mer
Ramelteon, TRC
Produktkod. 30137439
Change view
Kvantitet:
5 mg
25 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137439 500 mg
30135052 25 mg
30004679 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137439 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR110051500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 196597-26-9
Molekylformel C16 H21 N O2
Molekylvikt (g/mol) 259.34
Synonym N-[2-[(8S)-1,6,7,8-Tetrahydro-2H-indeno[5,4-b]furan-8-yl]ethyl]propanamide, Propanamide, N-[2-(1,6,7,8-tetrahydro-2H-indeno[5,4-b]furan-8-yl)ethyl]-, (S)-, (S)-N-[2-(1,6,7,8-Tetrahydro-2H-indeno-[5,4-b]furan-8-yl)ethyl]propionamide, (S)-N-[2-(1,6,7,8-Tetrahydro-2H-indeno[5,4-b]furan-8-yl)ethyl]propionamide, Ramelteon, Rozerem, TAK 375
IUPAC-namn N-[2-[(8S)-2,6,7,8-tetrahydro-1H-cyclopenta[e][1]benzofuran-8-yl]ethyl]propanamide
LEDER CCC(=O)NCC[C@@H]1CCc2ccc3OCCc3c12
Specifikationer
Kemiskt namn eller material Ramelteon
Kvantitet 500 mg
InChI formel InChI=1S/C16H21NO2/c1-2-15(18)17-9-7-12-4-3-11-5-6-14-13(16(11)12)8-10-19-14/h5-6,12H,2-4,7-10H2,1H3,(H,17,18)/t12-/m0/s1
Formel vikt 259.1572