Läs mer
Ramifenazone Hydrochloride Salt, TRC
Produktkod. 30417394
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30417394 100 mg
1 options
Produktkod. 30417394 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR110500100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ramifenazone HCl Salt
CAS 18342-39-7
Molekylformel C14 H19 N3 O . Cl H
Kvantitet 100 mg
Synonym 3H-Pyrazol-3-one','1,2-dihydro-1,5-dimethyl-4-[(1-methylethyl)amino]-2-phenyl-','monohydrochloride (9CI), Antipyrine','4-(isopropylamino)-','monohydrochloride (8CI), Isopyrine hydrochloride, 3H-Pyrazol-3-one','1,2-dihydro-1,5-dimethyl-4-[(1-methylethyl)amino]-2-phenyl-','hydrochloride (1:1), Ramifenazone Hydrochloride
InChI formel InChI=1S/C14H19N3O.ClH/c1-10(2)15-13-11(3)16(4)17(14(13)18)12-8-6-5-7-9-12;/h5-10,15H,1-4H3;1H
LEDER Cl.CC(C)NC1=C(C)N(C)N(C1=O)c2ccccc2
IUPAC-namn 1,5-dimethyl-2-phenyl-4-(propan-2-ylamino)pyrazol-3-one;hydrochloride
Molekylvikt (g/mol) 281.78
Formel vikt 281.1295