Läs mer
Ramipril Acyl-Beta-D-glucuronide, TRC
Produktkod. 30413113
Change view
Kvantitet:
1 mg
2 mg
5 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30413113 2 mg
30410606 1 mg
30418028 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30413113 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR1110052MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1357570-21-8
Molekylformel C29H40N2O11
Molekylvikt (g/mol) 592.63
Synonym 1-[(2S,3aS,6aS)-1-[(2S)-2-[[(1S)-1-(Ethoxycarbonyl)-3-phenylpropyl]amino]-1-oxopropyl]octahydrocyclopenta[b]pyrrole-2-carboxylate] β-D-Glucopyranuronic Acid
IUPAC-namn (2S,3S,4S,5R,6S)-6-[(2S,3aS,6aS)-1-[(2S)-2-[[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino]propanoyl]-3,3a,4,5,6,6a-hexahydro-2H-cyclopenta[b]pyrrole-2-carbonyl]oxy-3,4,5-trihydroxyoxane-2-carboxylic acid
LEDER C[C@@H](C(N1[C@](CCC2)([H])[C@]2([H])C[C@H]1C(O[C@@H]([C@@H]3O)O[C@H](C(O)=O)[C@@H](O)[C@@H]3O)=O)=O)N[C@H](C(OCC)=O)CCC4=CC=CC=C4
Specifikationer
Kemiskt namn eller material Ramipril Acyl-beta-D-glucuronide
Kvantitet 2 mg
InChI formel InChI=1S/C29H40N2O11/c1-3-40-27(38)18(13-12-16-8-5-4-6-9-16)30-15(2)25(35)31-19-11-7-10-17(19)14-20(31)28(39)42-29-23(34)21(32)22(33)24(41-29)26(36)37/h4-6,8-9,15,17-24,29-30,32-34H,3,7,10-14H2,1-2H3,(H,36,37)/t15-,17-,18-,19-,20-,21-,22-,23+,24-,29-/m0/s1
Formel vikt 592.2632