missing translation for 'onlineSavingsMsg'
Läs mer

Ramipril-d5, TRC

Product Code. 30416036
Change view
Click to view available options
Kvantitet:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30416036 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30416036 Supplier Toronto Research Chemicals Supplier No. TRCR1110021MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Ramipril-d5
CAS 1132661-86-9
Molekylformel C23H27D5N2O5
Kvantitet 1 mg
Synonym (2S,3aS,6aS)-1-[(2S)-2-[[(1S)-1-(Ethoxycarbonyl)-3-(phenyl-d5)propyl]amino]-1-oxopropyl]octahydrocyclopenta[b]pyrrole-2-carboxylic Acid, Altace-d5, Cardace-d5, Delix-d5, HOE-498-d5, Pramace-d5, Ramace-d5, Unipril-d5
InChI formel InChI=1S/C23H32N2O5/c1-3-30-23(29)18(13-12-16-8-5-4-6-9-16)24-15(2)21(26)25-19-11-7-10-17(19)14-20(25)22(27)28/h4-6,8-9,15,17-20,24H,3,7,10-14H2,1-2H3,(H,27,28)/t15-,17-,18-,19-,20-/m0/s1/i4D,5D,6D,8D,9D
LEDER C[C@@H](C(N1[C@](CCC2)([H])[C@]2([H])C[C@H]1C(O)=O)=O)N[C@H](C(OCC)=O)CCC3=C([2H])C([2H])=C([2H])C([2H])=C3[2H]
IUPAC-namn (2S,3aS,6aS)-1-[(2S)-2-[[(2S)-1-ethoxy-1-oxo-4-(2,3,4,5,6-pentadeuteriophenyl)butan-2-yl]amino]propanoyl]-3,3a,4,5,6,6a-hexahydro-2H-cyclopenta[b]pyrrole-2-carboxylic acid
Molekylvikt (g/mol) 421.54
Formel vikt 421.2625
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.