missing translation for 'onlineSavingsMsg'
Läs mer

Ramiprilat Acyl-Beta-D-glucuronide >65%, TRC

Produktkod. 30428526
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30428526 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30428526 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR1110201MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1357570-22-9
Molekylformel C27H36N2O11
Molekylvikt (g/mol) 564.58
Synonym Ramiprilat Acyl-B-D-Glucuronide
LEDER C[C@H](N[C@@H](CCc1ccccc1)C(=O)O)C(=O)N2[C@H]3CCC[C@H]3C[C@H]2C(=O)O[C@@H]4O[C@@H]([C@@H](O)[C@H](O)[C@H]4O)C(=O)O

Specifications

Kemiskt namn eller material Ramiprilat Acyl-beta-D-glucuronide
Kvantitet 1 mg
InChI formel InChi=1S/C27H36N2O11/c1-13(28-16(24(34)35)11-10-14-6-3-2-4-7-14)23(33)29-17-9-5-8-15(17)12-18(29)26(38)40-27-21(32)19(30)20(31)22(39-27)25(36)37/h2-4,6-7,13,15-22,27-28,30-32H,5,8-12H2,1H3,(H,34,35)(H,36,37)/t13-,15-,16-,17-,18-,19-,20-,21+,22-,27-/m0/s1
Formel vikt 564.2319
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.